siRNA Details
Gene Symbol: ITGAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000887.4 | NP_000878.2 | 28 | 3387 | ||
NM_001286375.1 | NP_001273304.1 | 28 | 3387 | |||
GenBank mRNA | AK074047.1 | - | 20 | 3131 | ||
BC038237.1 | - | 28 | 3299 | 5745308 | ||
M81695.1 | - | 28 | 3330 |
Gene Symbol: ITGAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318185.1 | NP_001305114.1 | 28 | 3303 | ||
NM_005353.2 | NP_005344.2 | 28 | 3300 | |||
XM_011545835.2 | XP_011544137.1 | 28 | 3464 | |||
XM_011545836.2 | XP_011544138.1 | 28 | 3460 | |||
XM_011545837.2 | XP_011544139.1 | 28 | 3463 | |||
XM_011545838.2 | XP_011544140.1 | 28 | 3430 | |||
XM_011545839.2 | XP_011544141.1 | 28 | 3412 | |||
XM_011545841.2 | XP_011544143.1 | 28 | 3372 | |||
XM_011545842.2 | XP_011544144.1 | 27 | 3205 | |||
XM_011545844.2 | XP_011544146.1 | 24 | 2889 | |||
XM_011545847.2 | XP_011544149.1 | 20 | 2227 | |||
XM_017023215.1 | XP_016878704.1 | 27 | 3388 | |||
GenBank mRNA | AB208945.1 | - | 28 | 3300 | ||
U37028.1 | - | 28 | 3254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
