siRNA Details
Gene Symbol: GLE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001499.2 | NP_001490.1 | 14 | 2086 | ||
GenBank mRNA | AF058922.1 | - | 14 | 2059 | ||
AK222842.1 | - | 14 | 2032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
