siRNA Details
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311818.1 | - | 4 | 767 | ||
M12937.1 | - | 4 | 587 | |||
X00318.1 | - | 4 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
