siRNA Details
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002216.2 | NP_002207.2 | 21 | 3003 | ||
GenBank mRNA | AK225277.1 | - | 23 | 2866 | ||
AK225329.1 | - | 21 | 2908 | |||
AK291379.1 | - | 21 | 2951 | |||
BC132685.1 | - | 21 | 2890 | 40146707 | ||
X07173.1 | - | 21 | 2904 |
