siRNA Details
Gene Symbol: SLC28A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287762.1 | NP_001274691.1 | 12 | 1310 | ||
NM_001321721.1 | NP_001308650.1 | 13 | 1426 | |||
NM_001321722.1 | NP_001308651.1 | 13 | 1426 | |||
NM_004213.4 | NP_004204.3 | 13 | 1426 | |||
XM_011522203.2 | XP_011520505.1 | 13 | 1409 | |||
XM_011522204.2 | XP_011520506.1 | 14 | 1929 | |||
XM_011522205.2 | XP_011520507.1 | 13 | 1813 | |||
XM_011522206.2 | XP_011520508.1 | 12 | 1612 | |||
XM_011522207.2 | XP_011520509.1 | 12 | 1254 | |||
XM_011522208.2 | XP_011520510.1 | 11 | 1531 | |||
XM_011522209.2 | XP_011520511.1 | 12 | 1331 | |||
XM_011522210.2 | XP_011520512.1 | 13 | 1409 | |||
XM_011522211.2 | XP_011520513.1 | 12 | 1644 | |||
XM_011522212.1 | XP_011520514.1 | 13 | 1409 | |||
XM_011522214.2 | XP_011520516.1 | 13 | 1409 | |||
XM_011522216.2 | XP_011520518.1 | 11 | 1165 | |||
XM_017022723.1 | XP_016878212.1 | 13 | 1409 | |||
GenBank mRNA | AF309632.1 | - | 13 | 1405 | ||
AK291997.1 | - | 13 | 1426 | |||
AK296074.1 | - | 13 | 1425 | |||
AK298369.1 | - | 13 | 1426 | |||
BC126204.1 | - | 12 | 1316 | 8991920 | ||
BC126206.1 | - | 12 | 1316 | 8991922 | ||
U62967.1 | - | 11 | 1218 | |||
U62968.1 | - | 11 | 1218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
