siRNA Details
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 3, 4 | 400 | ||
NR_110887.1 | 3, 4 | 400 | ||||
GenBank mRNA | AF001008.1 | - | 3, 4 | 299 | ||
AF001174.1 | - | 3, 4 | 305 | |||
AF031135.1 | - | 3, 4 | 314 | |||
AK308969.1 | - | 3, 4 | 316 | |||
AK309046.1 | - | 3, 4 | 502 | |||
BC027933.1 | - | 3, 4 | 357 | 5199628 | ||
BM857051.1 | - | 3, 4 | 384 | |||
BX361670.2 | - | 3, 4 | 384 | |||
BX444192.2 | - | 3, 4 | 399 | |||
CR456514.1 | - | 3, 4 | 301 | |||
U53442.1 | - | 3, 4 | 318 | |||
U92268.1 | - | 3, 4 | 303 | |||
Y14440.1 | - | 3, 4 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
