siRNA Details
Gene Symbol: GPR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097579.1 | NP_001091048.1 | 3 | 372 | ||
NM_005300.3 | NP_005291.1 | 3 | 296 | |||
XM_005272597.3 | XP_005272654.1 | 3 | 2799 | |||
GenBank mRNA | AF039686.1 | - | 1 | 166 | ||
AK027780.1 | - | 3 | 372 | |||
AK074627.1 | - | 3 | 518 | |||
AK075217.1 | - | 3 | 287 | |||
AK122945.1 | - | 1 | 1727 | |||
BC020678.1 | - | 3 | 263 | 4770479 | ||
DQ106398.1 | - | 3 | 363 | |||
DQ106399.1 | - | 4 | 430 | |||
DQ106400.1 | - | 4 | 476 | |||
DQ106401.1 | - | 3 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
