siRNA Details
Gene Symbol: INPP5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321042.1 | NP_001307971.1 | 6 | 1039 | ||
NM_005539.4 | NP_005530.3 | 9 | 986 | |||
XM_017016204.1 | XP_016871693.1 | 8 | 640 | |||
XM_017016205.1 | XP_016871694.1 | 9 | 981 | |||
GenBank mRNA | AF273055.1 | - | 9 | 929 | ||
BC052265.1 | - | 8 | 584 | |||
BC062300.1 | - | 8 | 584 | |||
BC096280.2 | - | 10 | 730 | 40006844 | ||
BC096281.2 | - | 9 | 649 | 40006846 | ||
X77567.1 | - | 9 | 767 | |||
Z31695.1 | - | 9 | 622 |
