siRNA Details
Gene Symbol: RAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002880.3 | NP_002871.1 | 3, 4 | 742 | ||
XM_005265355.2 | XP_005265412.1 | 3, 4 | 470 | |||
XM_005265357.2 | XP_005265414.1 | 3, 4 | 658 | |||
XM_011533974.2 | XP_011532276.1 | 4, 5 | 785 | |||
XM_017006966.1 | XP_016862455.1 | 3, 4 | 446 | |||
GenBank mRNA | AK226028.1 | - | 3, 4 | 658 | ||
AK312248.1 | - | 3, 4 | 636 | |||
BC018119.2 | - | 3, 4 | 656 | 3904404 | ||
KU178297.1 | - | 2, 3 | 327 | |||
KU178298.1 | - | 2, 3 | 327 | |||
KU178299.1 | - | 2, 3 | 327 | |||
X03484.1 | - | 3, 4 | 456 |
Gene Symbol: RAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051846.1 | - | 1416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
