siRNA Details
Gene Symbol: RECQL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003715.3 | NP_001003715.1 | 9 | 1502 | ||
NM_001003716.3 | NP_001003716.1 | 8 | 1969 | |||
XM_017025344.1 | XP_016880833.1 | 8 | 1982 | |||
GenBank mRNA | AB042823.1 | - | 8 | 1974 | ||
AB042825.1 | - | 9 | 1502 | |||
AK075084.1 | - | 9 | 1493 | |||
BC016911.1 | - | 9 | 1678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
