siRNA Details
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000082.3 | NP_000073.1 | 8 | 739 | ||
NM_001007233.2 | NP_001007234.1 | 9 | 957 | |||
NM_001290285.1 | NP_001277214.1 | 7 | 657 | |||
GenBank mRNA | AK056931.1 | - | 9 | 930 | ||
AK226129.1 | - | 5 | 516 | |||
AK294856.1 | - | 7 | 638 | |||
AK304610.1 | - | 8 | 669 | |||
AK314511.1 | - | 8 | 712 | |||
BT020021.1 | - | 8 | 669 | |||
CR536563.1 | - | 8 | 669 | |||
U28413.1 | - | 8 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
