siRNA Details
Gene Symbol: CALCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000728.3 | NP_000719.1 | 4 | 413 | ||
GenBank mRNA | AK054728.1 | - | 9 | 1635 | ||
AK291188.1 | - | 4 | 416 | |||
BC092468.1 | - | 4 | 417 | 5313089 | ||
BI668461.1 | - | 4 | 422 | |||
X02404.1 | - | 2 | 177 |
Gene Symbol: CALCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033953.2 | NP_001029125.1 | 4 | 418 | ||
NR_125898.1 | 5 | 566 | ||||
XM_017018283.1 | XP_016873772.1 | 4 | 1612 | |||
XM_017018284.1 | XP_016873773.1 | 4 | 1636 | |||
GenBank mRNA | BX094781.1 | - | 1 | 88 | ||
CB048118.1 | - | 1 | 100 | |||
K03512.1 | - | 2 | 202 | |||
M64486.1 | - | 5 | 554 | |||
X02330.1 | - | 2 | 1341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
