siRNA Details
Gene Symbol: EPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000799.2 | NP_000790.2 | 3 | 401 | ||
GenBank mRNA | BC093628.1 | - | 3 | 298 | 7939473 | |
BC111937.1 | - | 3 | 298 | 8327405 | ||
BC143225.1 | - | 3 | 298 | 9051726 | ||
JN849371.1 | - | 3 | 220 | |||
KF178447.1 | - | 2 | 229 | |||
S65458.1 | - | 1 | 51 | |||
X02157.1 | - | 3 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
