siRNA Details
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160124.1 | NP_001153596.1 | 4 | 1322 | ||
NM_001160125.1 | NP_001153597.1 | 3 | 1324 | |||
NM_001300.5 | NP_001291.3 | 4 | 1448 | |||
NR_027653.1 | 4 | 1294 | ||||
GenBank mRNA | AB017493.1 | - | 4 | 1302 | ||
AF001461.1 | - | 4 | 1210 | |||
BC000311.2 | - | 4 | 1367 | 2821009 | ||
BC004301.1 | - | 4 | 1365 | |||
BX537374.1 | - | 2 | 1806 | |||
U44975.1 | - | 4 | 1206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
