siRNA Details
Gene Symbol: CSTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326.2 | NP_001317.1 | 8 | 673 | ||
GenBank mRNA | AK290836.1 | - | 8 | 673 | ||
AK294204.1 | - | 5 | 266 | |||
AK297371.1 | - | 7 | 555 | |||
BC108319.1 | - | 8 | 647 | 6197136 | ||
U15782.1 | - | 8 | 638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
