siRNA Details
Gene Symbol: ERN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001433.3 | NP_001424.3 | 6 | 484 | ||
XM_017024347.1 | XP_016879836.1 | 6 | 1838 | |||
XM_017024348.1 | XP_016879837.1 | 6 | 1838 | |||
GenBank mRNA | AB209869.1 | - | 6 | 467 | ||
AF059198.1 | - | 6 | 467 | |||
AK292403.1 | - | 6 | 510 | |||
BC130405.1 | - | 6 | 417 | 40146436 | ||
BC130407.1 | - | 6 | 417 | 40146438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
