siRNA Details
Gene Symbol: CCT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001762.3 | NP_001753.1 | 3 | 487 | ||
GenBank mRNA | AB063318.1 | - | 3 | 459 | ||
AB209728.1 | - | 3 | 319 | |||
AF385084.1 | - | 3 | 378 | |||
AK297751.1 | - | 2 | 172 | |||
AK298371.1 | - | 4 | 252 | |||
AK313819.1 | - | 3 | 345 | |||
AK316230.1 | - | 4 | 305 | |||
AM295155.1 | - | 3 | 298 | |||
BC106941.1 | - | 3 | 403 | |||
BC106942.1 | - | 3 | 403 | 40034021 | ||
BI464886.1 | - | 3 | 521 | |||
L27706.1 | - | 3 | 409 | |||
M94083.1 | - | 1 | 58 |
Gene Symbol: CCT6P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033416.1 | 2 | 334 | |||
GenBank mRNA | BX649060.1 | - | 2 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
