siRNA Details
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004435.2 | NP_004426.2 | 3 | 811 | ||
XM_011518347.2 | XP_011516649.1 | 4 | 1036 | |||
GenBank mRNA | AK130973.1 | - | 3 | 292 | ||
BC004922.2 | - | 3 | 686 | 3604703 | ||
BC016351.1 | - | 3 | 841 | 4080883 | ||
X79444.1 | - | 3 | 792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
