siRNA Details
Gene Symbol: RUNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031680.2 | NP_001026850.1 | 6 | 3377 | ||
NM_001320672.1 | NP_001307601.1 | 7 | 3210 | |||
NM_004350.2 | NP_004341.1 | 5 | 3307 | |||
XM_005246024.4 | XP_005246081.1 | 7 | 3085 | |||
XM_011542351.1 | XP_011540653.1 | 5 | 3218 | |||
GenBank mRNA | Z35278.1 | - | 5 | 2902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
