siRNA Details
Gene Symbol: CD74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025159.2 | NP_001020330.1 | 5 | 699 | ||
NM_004355.3 | NP_004346.1 | 5 | 699 | |||
XM_017010089.1 | XP_016865578.1 | 5 | 675 | |||
XM_017010090.1 | XP_016865579.1 | 5 | 675 | |||
GenBank mRNA | AK292076.1 | - | 5 | 514 | ||
AK297889.1 | - | 5 | 514 | |||
AK300669.1 | - | 4 | 250 | |||
BC018726.1 | - | 5 | 523 | 4853578 | ||
BT019505.1 | - | 5 | 512 | |||
K01144.1 | - | 5 | 563 | |||
X00497.1 | - | 5 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
