siRNA Details
Gene Symbol: MAP3K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004672.4 | NP_004663.3 | 2, 3 | 760 | ||
XM_017002771.1 | XP_016858260.1 | 1 | 129 | |||
XM_017002772.1 | XP_016858261.1 | 2, 3 | 1931 | |||
GenBank mRNA | AB167411.1 | - | 2, 3 | 491 | ||
AI582755.1 | - | 2, 3 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
