siRNA Details
Gene Symbol: DSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004415.3 | NP_004406.2 | 23 | 4420 | ||
GenBank mRNA | J05211.1 | - | 1 | 722 | ||
M77830.3 | - | 23 | 4358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
