siRNA Details
Gene Symbol: FUT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004479.3 | NP_004470.1 | 2 | 1859 | ||
GenBank mRNA | AK313124.1 | - | 2 | 1493 | ||
BC074746.2 | - | 2 | 1035 | 30915206 | ||
BC086312.1 | - | 2 | 1859 | 6651145 | ||
U08112.1 | - | 2 | 1144 | |||
U11282.1 | - | 2 | 1147 | |||
X78031.1 | - | 2 | 1132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
