siRNA Details
Gene Symbol: FUT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004480.4 | NP_004471.4 | 3 | 1581 | ||
NM_178155.2 | NP_835368.1 | 4 | 2011 | |||
NM_178156.2 | NP_835369.1 | 3 | 1913 | |||
NR_038167.1 | 4 | 2011 | ||||
NR_038170.1 | 4 | 1094 | ||||
XM_017021136.1 | XP_016876625.1 | 4 | 1094 | |||
XM_017021137.1 | XP_016876626.1 | 4 | 738 | |||
XM_017021138.1 | XP_016876627.1 | 3 | 605 | |||
XM_017021139.1 | XP_016876628.1 | 3 | 996 | |||
XM_017021140.1 | XP_016876629.1 | 2 | 2946 | |||
GenBank mRNA | AB049740.2 | - | 2 | 512 | ||
AJ536053.1 | - | 4 | 1479 | |||
AJ536054.1 | - | 4 | 1479 | |||
AJ536055.1 | - | 3 | 1381 | |||
AJ536056.1 | - | 4 | 2009 | |||
AJ539535.1 | - | 3 | 609 | |||
AJ539536.1 | - | 3 | 609 | |||
AK292413.1 | - | 4 | 732 | |||
AK294242.1 | - | 3 | 299 | |||
BC025385.2 | - | 3 | 386 | |||
BC142958.1 | - | 4 | 738 | 8860361 | ||
D89289.1 | - | 2 | 426 | |||
Y17976.1 | - | 4 | 712 | |||
Y17977.1 | - | 4 | 712 | |||
Y17978.1 | - | 4 | 1000 | |||
Y17979.1 | - | 4 | 1000 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016893, NM_001252614, BC010666, AK051811, AK163494, and others.
Gene Ontology Categories:
