siRNA Details
Gene Symbol: DMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290321.2 | NP_001277250.1 | 11 | 1853 | ||
NM_001290322.2 | NP_001277251.1 | 11 | 3161 | |||
NM_005509.5 | NP_005500.4 | 11 | 1853 | |||
XM_005271909.4 | XP_005271966.1 | 12 | 1551 | |||
XM_005271910.4 | XP_005271967.1 | 11 | 1896 | |||
XM_005271912.2 | XP_005271969.1 | 12 | 1535 | |||
XM_011543212.2 | XP_011541514.1 | 11 | 1896 | |||
XM_011543213.2 | XP_011541515.1 | 12 | 1481 | |||
XM_011543214.2 | XP_011541516.1 | 8 | 1062 | |||
XM_011543215.2 | XP_011541517.1 | 7 | 898 | |||
XM_017009141.1 | XP_016864630.1 | 12 | 1551 | |||
XM_017009142.1 | XP_016864631.1 | 12 | 1481 | |||
XM_017009143.1 | XP_016864632.1 | 13 | 1730 | |||
XM_017009144.1 | XP_016864633.1 | 11 | 1433 | |||
XM_017009145.1 | XP_016864634.1 | 11 | 1303 | |||
XM_017009146.1 | XP_016864635.1 | 11 | 2745 | |||
XM_017009147.1 | XP_016864636.1 | 11 | 1896 | |||
XM_017009148.1 | XP_016864637.1 | 10 | 1699 | |||
GenBank mRNA | AB593142.1 | - | 11 | 3164 | ||
AJ005821.1 | - | 11 | 1450 | |||
BC028004.1 | - | 12 | 1438 | |||
BC144537.1 | - | 11 | 1396 | 9053064 | ||
BC150616.1 | - | 11 | 1396 | 9056986 |
