siRNA Details
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 7 | 1392 | ||
NR_135748.1 | 6 | 1441 | ||||
NR_135749.1 | 6 | 1421 | ||||
NR_135750.1 | 7 | 1392 | ||||
NR_135751.1 | 6 | 1281 | ||||
NR_135752.1 | 5 | 1048 | ||||
XM_005254150.3 | XP_005254207.1 | 6 | 870 | |||
XM_011521194.1 | XP_011519496.1 | 7 | 1480 | |||
XM_011521195.2 | XP_011519497.1 | 7 | 1219 | |||
XM_011521196.1 | XP_011519498.1 | 7 | 1145 | |||
XM_011521197.2 | XP_011519499.1 | 6 | 979 | |||
XM_011521198.1 | XP_011519500.1 | 7 | 1664 | |||
XM_011521199.2 | XP_011519501.2 | 7 | 1436 | |||
XM_017021902.1 | XP_016877391.1 | 7 | 1620 | |||
GenBank mRNA | AK094322.1 | - | 7 | 970 | ||
AK293685.1 | - | 6 | 662 | |||
AK293790.1 | - | 6 | 1248 | |||
AK293939.1 | - | 6 | 1045 | |||
AK293961.1 | - | 5 | 830 | |||
AK294063.1 | - | 6 | 731 | |||
AK294735.1 | - | 7 | 863 | |||
AK294908.1 | - | 6 | 1280 | |||
AK296271.1 | - | 5 | 481 | |||
AK296472.1 | - | 6 | 883 | |||
AK308770.1 | - | 5 | 753 | |||
BC108652.1 | - | 6 | 915 | 6190035 | ||
BC121001.1 | - | 6 | 979 | 40116549 | ||
BC121002.1 | - | 6 | 979 | 40116552 |
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 7 | 1295 | ||
NM_001321926.1 | NP_001308855.1 | 6 | 1163 | |||
NM_001321927.1 | NP_001308856.1 | 7 | 1388 | |||
NM_001321928.1 | NP_001308857.1 | 7 | 1149 | |||
NM_001321929.1 | NP_001308858.1 | 6 | 1147 | |||
NR_135856.1 | 6 | 1394 | ||||
XM_005254498.3 | XP_005254555.1 | 6 | 870 | |||
XM_017022369.1 | XP_016877858.1 | 7 | 1220 | |||
XM_017022370.1 | XP_016877859.1 | 7 | 1145 | |||
GenBank mRNA | AK223365.1 | - | 6 | 975 | ||
AK295776.1 | - | 6 | 1039 | |||
AK316052.1 | - | 7 | 795 | |||
AK316124.1 | - | 7 | 1246 | |||
AK316319.1 | - | 7 | 1149 | |||
BC001926.1 | - | 6 | 980 | 2820642 | ||
BC006467.1 | - | 6 | 906 | 3535486 | ||
BT006628.1 | - | 6 | 803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
