siRNA Details
Gene Symbol: DLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021120.3 | NP_066943.2 | 6 | 1303 | ||
XM_006724625.2 | XP_006724688.1 | 6 | 1308 | |||
XM_006724626.2 | XP_006724689.1 | 6 | 1308 | |||
XM_011530883.1 | XP_011529185.1 | 6 | 1308 | |||
GenBank mRNA | AB209328.1 | - | 6 | 1303 | ||
AK315041.1 | - | 6 | 1242 | |||
BC093864.1 | - | 6 | 1004 | 7939709 | ||
BC093866.1 | - | 6 | 1004 | 7939711 | ||
U49089.1 | - | 6 | 1189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
