siRNA Details
Gene Symbol: GRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001619.3 | NP_001610.2 | 3 | 508 | ||
XM_011544773.1 | XP_011543075.1 | 3 | 249 | |||
GenBank mRNA | AB209588.1 | - | 2 | 864 | ||
BC037963.1 | - | 3 | 487 | 5585846 | ||
BC090863.1 | - | 3 | 476 | 6203199 | ||
KU177896.1 | - | 3 | 242 | |||
KU177897.1 | - | 3 | 242 | |||
M80776.1 | - | 3 | 257 | |||
X61157.1 | - | 3 | 349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
