siRNA Details
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004922.2 | - | 3 | 1013 | 3604703 | |
X79444.1 | - | 3 | 1119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
