siRNA Details
Gene Symbol: GRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004106.2 | NP_001004106.1 | 16 | 2180 | ||
NM_002082.3 | NP_002073.2 | 16 | 2182 | |||
XM_006714858.3 | XP_006714921.1 | 14 | 2823 | |||
XM_006714859.2 | XP_006714922.1 | 16 | 2094 | |||
XM_011534537.2 | XP_011532839.1 | 15 | 1934 | |||
XM_011534538.1 | XP_011532840.1 | 9 | 1376 | |||
GenBank mRNA | AF040751.1 | - | 16 | 2024 | ||
BC009277.2 | - | 16 | 2120 | 4053197 | ||
L16862.1 | - | 16 | 2082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
