siRNA Details
Gene Symbol: ALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004304.4 | NP_004295.2 | 4, 5 | 2109 | ||
GenBank mRNA | AB209477.4 | - | 4, 5 | 1646 | ||
EU788003.1 | - | 4, 5 | 2064 | |||
EU788004.1 | - | 4, 5 | 2064 | |||
EU788005.1 | - | 4, 5 | 2064 | |||
EU788006.1 | - | 4, 5 | 2064 | |||
EU788007.1 | - | 4, 5 | 2064 | |||
EU788008.1 | - | 4, 5 | 2064 | |||
EU788009.1 | - | 4, 5 | 2064 | |||
EU788010.1 | - | 4, 5 | 2064 | |||
FJ838786.1 | - | 4, 5 | 2064 | |||
FJ838787.1 | - | 4, 5 | 2064 | |||
GU128152.1 | - | 4, 5 | 1157 | |||
GU128153.1 | - | 4, 5 | 1157 | |||
GU128154.1 | - | 4, 5 | 1157 | |||
GU128155.1 | - | 4, 5 | 1157 | |||
GU128156.1 | - | 4, 5 | 1157 | |||
GU128157.1 | - | 4, 5 | 1157 | |||
U62540.1 | - | 4, 5 | 2068 | |||
U66559.1 | - | 4, 5 | 1305 |
