siRNA Details
Gene Symbol: GRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005160.3 | NP_005151.2 | 5 | 533 | ||
XM_011529975.2 | XP_011528277.1 | 3 | 155 | |||
GenBank mRNA | AK058103.1 | - | 4 | 323 | ||
AK292234.1 | - | 5 | 399 | |||
AK310398.1 | - | 5 | 401 | |||
BC036797.1 | - | 6 | 477 | 5590378 | ||
BX387339.2 | - | 5 | 578 | |||
X69117.1 | - | 5 | 386 |
