siRNA Details
Gene Symbol: GRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005160.3 | NP_005151.2 | 17, 18 | 1638 | ||
XM_011529975.2 | XP_011528277.1 | 16, 17 | 1348 | |||
GenBank mRNA | AK058103.1 | - | 16, 17 | 1428 | ||
AK292234.1 | - | 17, 18 | 1504 | |||
BC036797.1 | - | 19, 20 | 1732 | 5590378 | ||
X69117.1 | - | 17, 18 | 1491 |
