siRNA Details
Gene Symbol: FUT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004480.4 | NP_004471.4 | 4 | 1635 | ||
NM_178155.2 | NP_835368.1 | 5 | 2065 | |||
NM_178156.2 | NP_835369.1 | 4 | 1967 | |||
NR_038167.1 | 5 | 2065 | ||||
NR_038170.1 | 5 | 1148 | ||||
XM_017021136.1 | XP_016876625.1 | 5 | 1148 | |||
XM_017021137.1 | XP_016876626.1 | 5 | 792 | |||
XM_017021138.1 | XP_016876627.1 | 4 | 659 | |||
XM_017021139.1 | XP_016876628.1 | 4 | 1050 | |||
XM_017021140.1 | XP_016876629.1 | 3 | 3000 | |||
GenBank mRNA | AB049740.2 | - | 3 | 566 | ||
AJ536053.1 | - | 5 | 1533 | |||
AJ536054.1 | - | 5 | 1533 | |||
AJ536055.1 | - | 4 | 1435 | |||
AJ536056.1 | - | 5 | 2063 | |||
AJ539535.1 | - | 4 | 663 | |||
AJ539536.1 | - | 4 | 663 | |||
AK294242.1 | - | 4 | 353 | |||
BC025385.2 | - | 4 | 440 | |||
BC142958.1 | - | 5 | 792 | 8860361 | ||
D89289.1 | - | 3 | 480 | |||
Y17976.1 | - | 5 | 766 | |||
Y17977.1 | - | 5 | 766 | |||
Y17978.1 | - | 5 | 1054 | |||
Y17979.1 | - | 5 | 1054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
