siRNA Details
Gene Symbol: GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291866.1 | NP_001278795.1 | 3 | 421 | ||
NM_004481.4 | NP_004472.1 | 3 | 343 | |||
XM_017000963.1 | XP_016856452.1 | 3 | 303 | |||
XM_017000964.1 | XP_016856453.1 | 3 | 928 | |||
XM_017000965.1 | XP_016856454.1 | 3 | 421 | |||
XM_017000966.1 | XP_016856455.1 | 3 | 246 | |||
GenBank mRNA | AK290048.1 | - | 3 | 269 | ||
AK300453.1 | - | 3 | 421 | |||
AK304029.1 | - | 2 | 173 | |||
BC041120.1 | - | 3 | 329 | 5553465 | ||
BG773173.1 | - | 3 | 349 | |||
X85019.1 | - | 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
