siRNA Details
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 11, 12 | 2304 | ||
GenBank mRNA | BC025289.1 | - | 10, 11 | 1738 |
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037162.1 | 10, 11 | 2477 | |||
GenBank mRNA | AF078842.1 | - | 6, 7 | 1129 | ||
AK315140.1 | - | 8, 9 | 1413 | |||
AL096725.1 | - | 4, 5 | 1802 | |||
BC098361.1 | - | 4, 5 | 1270 | |||
BC099735.1 | - | 4, 5 | 1270 | |||
BC105638.1 | - | 4, 5 | 1270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
