siRNA Details
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004522.2 | NP_004513.1 | 2 | 586 | ||
XM_017004062.1 | XP_016859551.1 | 2 | 577 | |||
GenBank mRNA | AB011103.1 | - | 2 | 559 | ||
BC002721.2 | - | 2 | 568 | |||
BC017298.2 | - | 2 | 541 | |||
BC025961.1 | - | 2 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
