siRNA Details
Gene Symbol: SLC41A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008487.1 | NP_001008487.1 | 10 | 1927 | ||
NM_001164475.1 | NP_001157947.1 | 10 | 1629 | |||
NM_017836.3 | NP_060306.3 | 11 | 2008 | |||
XM_005247564.1 | XP_005247621.1 | 11 | 1886 | |||
XM_017006706.1 | XP_016862195.1 | 11 | 1902 | |||
XM_017006707.1 | XP_016862196.1 | 11 | 1925 | |||
XM_017006708.1 | XP_016862197.1 | 10 | 1733 | |||
XM_017006709.1 | XP_016862198.1 | 10 | 1788 | |||
GenBank mRNA | AK021925.1 | - | 11 | 1694 | ||
AK091671.1 | - | 12 | 1812 | |||
AK093380.1 | - | 10 | 1924 | |||
AK094051.1 | - | 6 | 2017 | |||
BC009444.1 | - | 11 | 1826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
