siRNA Details
Gene Symbol: SH3TC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318480.1 | NP_001305409.1 | 17 | 3976 | ||
NM_018986.4 | NP_061859.3 | 17 | 3893 | |||
NR_134639.1 | 14 | 3456 | ||||
XM_006713889.2 | XP_006713952.1 | 14 | 3311 | |||
XM_006713891.3 | XP_006713954.1 | 11 | 2953 | |||
XM_011513488.1 | XP_011511790.1 | 17 | 3870 | |||
XM_011513489.2 | XP_011511791.1 | 15 | 3493 | |||
XM_017008300.1 | XP_016863789.1 | 17 | 4002 | |||
XM_017008301.1 | XP_016863790.1 | 17 | 3685 | |||
XM_017008302.1 | XP_016863791.1 | 14 | 3395 | |||
GenBank mRNA | AK074093.1 | - | 15 | 5732 | ||
AK074402.1 | - | 17 | 3770 | |||
AK093562.1 | - | 6 | 1814 | |||
AK126165.1 | - | 17 | 3849 | |||
AK128258.1 | - | 3 | 3354 | |||
AK222865.1 | - | 13 | 3281 |
