siRNA Details
Gene Symbol: SDR39U1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290292.1 | NP_001277221.1 | 5 | 1000 | ||
NM_001290293.2 | NP_001277222.1 | 5 | 1042 | |||
NM_001308075.1 | NP_001295004.1 | 5 | 962 | |||
NM_020195.2 | NP_064580.2 | 6 | 1082 | |||
XM_011536975.1 | XP_011535277.1 | 6 | 1126 | |||
XM_011536976.1 | XP_011535278.1 | 3 | 1130 | |||
GenBank mRNA | AF226050.1 | - | 5 | 1356 | ||
AK123513.1 | - | 4 | 2189 | |||
BC000989.2 | - | 6 | 1082 | 2900140 | ||
BC008566.1 | - | 5 | 1254 | |||
BX161384.1 | - | 4 | 993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
