siRNA Details
Gene Symbol: ARRDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020801.2 | NP_065852.1 | 3 | 666 | ||
GenBank mRNA | AB037797.1 | - | 3 | 638 | ||
AK291753.1 | - | 3 | 641 | |||
BC015928.2 | - | 3 | 633 | 3908916 | ||
BC053619.1 | - | 3 | 638 | 6155815 |
