siRNA Details
Gene Symbol: EXOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021807.3 | NP_068579.3 | 18 | 3360 | ||
XM_017012494.1 | XP_016867983.1 | 10 | 2210 | |||
GenBank mRNA | AB051486.1 | - | 18 | 3302 | ||
AF132734.1 | - | 12 | 2316 | |||
AF380839.1 | - | 18 | 3357 | |||
AK022751.1 | - | 11 | 2128 | |||
AK027688.1 | - | 18 | 3354 | |||
AK095495.1 | - | 10 | 2211 | |||
AL831989.2 | - | 18 | 3355 | |||
AL834475.1 | - | 15 | 2688 | |||
BC067263.1 | - | 18 | 3337 | 6183004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
