siRNA Details
Gene Symbol: FAM214B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317991.1 | NP_001304920.1 | 7, 8 | 1610 | ||
NM_025182.3 | NP_079458.2 | 7, 8 | 1611 | |||
NR_134455.1 | 8, 9 | 1247 | ||||
XM_005251588.1 | XP_005251645.1 | 7, 8 | 1745 | |||
XM_005251590.1 | XP_005251647.1 | 7, 8 | 1548 | |||
XM_005251591.1 | XP_005251648.1 | 7, 8 | 1584 | |||
XM_005251592.1 | XP_005251649.1 | 7, 8 | 1025 | |||
XM_005251593.1 | XP_005251650.1 | 7, 8 | 907 | |||
XM_005251594.1 | XP_005251651.1 | 7, 8 | 781 | |||
XM_005251596.4 | XP_005251653.1 | 7, 8 | 769 | |||
XM_005251597.4 | XP_005251654.1 | 7, 8 | 812 | |||
XM_005251598.4 | XP_005251655.1 | 7, 8 | 747 | |||
XM_011518037.1 | XP_011516339.1 | 7, 8 | 1567 | |||
XM_011518040.1 | XP_011516342.1 | 8, 9 | 1631 | |||
XM_011518043.1 | XP_011516345.1 | 8, 9 | 1049 | |||
XM_011518044.1 | XP_011516346.1 | 8, 9 | 969 | |||
XM_017015169.1 | XP_016870658.1 | 8, 9 | 1574 | |||
XM_017015170.1 | XP_016870659.1 | 8, 9 | 1265 | |||
GenBank mRNA | AB040972.1 | - | 6, 7 | 4341 | ||
AK021622.1 | - | 7, 8 | 1712 | |||
AK022837.1 | - | 8, 9 | 1125 | |||
AL583909.1 | - | 7, 8 | 1610 | |||
BC004406.1 | - | 7, 8 | 1596 | 3638496 |
