siRNA Details
Gene Symbol: ZNF541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277075.1 | NP_001264004.1 | 5, 6 | 2630 | ||
XM_005259311.4 | XP_005259368.1 | 6, 7 | 3570 | |||
XM_005259314.3 | XP_005259371.1 | 4, 5 | 1656 | |||
XM_011527367.2 | XP_011525669.1 | 6, 7 | 3585 | |||
XM_011527368.2 | XP_011525670.1 | 7, 8 | 2923 | |||
XM_011527369.2 | XP_011525671.1 | 7, 8 | 2927 | |||
XM_011527370.2 | XP_011525672.1 | 6, 7 | 2784 | |||
XM_011527371.2 | XP_011525673.1 | 6, 7 | 2771 | |||
XM_011527372.2 | XP_011525674.1 | 6, 7 | 3585 | |||
XM_011527374.2 | XP_011525676.1 | 6, 7 | 3420 | |||
XM_011527375.1 | XP_011525677.1 | 6, 7 | 3585 | |||
XM_017027351.1 | XP_016882840.1 | 6, 7 | 3585 | |||
GenBank mRNA | AL136846.1 | - | 3, 4 | 999 | ||
AL833862.1 | - | 3, 4 | 1401 | |||
BC101050.2 | - | 3, 4 | 996 | 40008070 | ||
BC101051.2 | - | 3, 4 | 996 | 40008071 | ||
BC101052.1 | - | 3, 4 | 996 | 40008072 | ||
BC101053.2 | - | 3, 4 | 996 | 40008073 |
