siRNA Details
Gene Symbol: ZNF541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277075.1 | NP_001264004.1 | 15 | 4439 | ||
XM_005259311.4 | XP_005259368.1 | 16 | 5379 | |||
XM_005259314.3 | XP_005259371.1 | 14 | 3465 | |||
XM_011527367.2 | XP_011525669.1 | 16 | 5394 | |||
XM_011527368.2 | XP_011525670.1 | 17 | 4732 | |||
XM_011527369.2 | XP_011525671.1 | 17 | 4736 | |||
XM_011527370.2 | XP_011525672.1 | 16 | 4593 | |||
XM_011527371.2 | XP_011525673.1 | 16 | 4580 | |||
XM_011527372.2 | XP_011525674.1 | 14 | 5229 | |||
XM_011527373.2 | XP_011525675.1 | 15 | 5220 | |||
XM_011527374.2 | XP_011525676.1 | 16 | 5229 | |||
XM_017027351.1 | XP_016882840.1 | 15 | 5253 | |||
GenBank mRNA | AL136846.1 | - | 13 | 2808 | ||
AL833862.1 | - | 7 | 2436 |
