siRNA Details
Gene Symbol: ZNF570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300993.2 | NP_001287922.1 | 6 | 817 | ||
NM_001321991.1 | NP_001308920.1 | 5 | 666 | |||
NM_001321992.1 | NP_001308921.1 | 4 | 997 | |||
NM_001321993.1 | NP_001308922.1 | 5 | 1140 | |||
NM_001321994.1 | NP_001308923.1 | 4 | 1056 | |||
NM_144694.3 | NP_653295.1 | 5 | 1081 | |||
XM_011526544.2 | XP_011524846.1 | 5 | 664 | |||
XM_011526545.2 | XP_011524847.1 | 4 | 568 | |||
GenBank mRNA | AK055353.1 | - | 5 | 1059 | ||
AK297561.1 | - | 4 | 758 | |||
AK308252.1 | - | 6 | 817 | |||
BC130425.1 | - | 5 | 964 | 40146456 | ||
BC130433.1 | - | 5 | 964 | 40146464 |
