siRNA Details
Gene Symbol: ZNF573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172689.1 | NP_001166160.1 | 6 | 1785 | ||
NM_001172690.1 | NP_001166161.1 | 5 | 1594 | |||
NM_001172691.1 | NP_001166162.1 | 5 | 1588 | |||
NM_001172692.1 | NP_001166163.1 | 4 | 1461 | |||
NM_152360.3 | NP_689573.3 | 8 | 1853 | |||
XM_011526446.2 | XP_011524748.1 | 6 | 1672 | |||
XM_017026275.1 | XP_016881764.1 | 6 | 1779 | |||
XM_017026276.1 | XP_016881765.1 | 6 | 2501 | |||
XM_017026277.1 | XP_016881766.1 | 6 | 1666 | |||
XM_017026278.1 | XP_016881767.1 | 4 | 1539 | |||
XM_017026279.1 | XP_016881768.1 | 4 | 1484 | |||
XM_017026280.1 | XP_016881769.1 | 5 | 1730 | |||
XM_017026281.1 | XP_016881770.1 | 4 | 2323 | |||
GenBank mRNA | AK055483.1 | - | 3 | 1603 | ||
AK074539.1 | - | 6 | 1781 | |||
BC015418.1 | - | 1 | 840 | |||
BC042170.2 | - | 4 | 1548 | 5014852 | ||
BC051263.1 | - | 1 | 415 | |||
BC064962.1 | - | 8 | 1823 | 6141400 |
