siRNA Details
Gene Symbol: RASGEF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF449764.1 | - | 13 | 2303 | ||
AK093160.1 | - | 11 | 1968 | |||
BC036802.1 | - | 15 | 2134 | |||
BC057759.1 | - | 12 | 1808 | 5268021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
