siRNA Details
Gene Symbol: OPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181744.3 | NP_859528.1 | 2 | 235 | ||
XM_017010410.1 | XP_016865899.1 | 1 | 361 | |||
XM_017010411.1 | XP_016865900.1 | 1 | 343 | |||
XM_017010416.1 | XP_016865905.1 | 2 | 237 | |||
GenBank mRNA | AY288419.1 | - | 2 | 207 | ||
AY377391.1 | - | 2 | 212 | |||
BC126194.1 | - | 2 | 235 | 8991910 | ||
BC126198.1 | - | 2 | 235 | 8991914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
