siRNA Details
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 2, 3 | 590 | ||
NM_001030313.2 | NP_001025484.1 | 2, 3 | 590 | |||
NM_201548.4 | NP_963842.1 | 2, 3 | 590 | |||
NR_027690.1 | 2, 3 | 590 | ||||
GenBank mRNA | AJ640141.1 | - | 2, 3 | 507 | ||
AJ697855.1 | - | 2, 3 | 489 | |||
AJ697856.1 | - | 2, 3 | 489 | |||
AY357073.1 | - | 2, 3 | 489 | |||
AY690329.1 | - | 2, 3 | 489 | |||
AY690331.1 | - | 2, 3 | 489 | |||
AY690333.1 | - | 2, 3 | 489 | |||
BC137498.1 | - | 2, 3 | 516 | 9021498 | ||
BC137499.1 | - | 2, 3 | 516 | 9021499 | ||
HQ426666.1 | - | 2, 3 | 396 | |||
HQ426667.1 | - | 2, 3 | 396 | |||
HQ426669.1 | - | 2, 3 | 396 | |||
HQ426671.1 | - | 2, 3 | 318 | |||
HQ426672.1 | - | 2, 3 | 318 | |||
HQ426673.1 | - | 2, 3 | 318 | |||
HQ426675.1 | - | 2, 3 | 318 | |||
HQ426678.1 | - | 3, 4 | 643 | |||
HQ426679.1 | - | 1, 2 | 251 | |||
JF290420.1 | - | 2, 3 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
